Proteintech
Manufacturer Code:125911AP150UL
Catalog # 50-172-8092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AKAP7 Fusion Protein Ag3297 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 7; A kinase anchor protein 18 kDa AKAP 18 AKAP 7 isoforms alpha and beta AKAP18 AKAP7; A-kinase anchor protein 18; A-kinase anchor protein 18 kDa; A-kinase anchor protein 18 {ECO:0000250|UniProtKB:F8VQ58}; A-kinase anchor protein 7; A-kinase anchor protein 7 isoform alpha; A-kinase anchor protein 7 isoform gamma; A-kinase anchor protein 7 isoforms alpha and beta; A-kinase anchor protein 7 isoforms delta and gamma; A-kinase anchor protein 9 kDa; A-kinase anchoring protein 18 ,isoform delta; AKAP 18; AKAP 9; AKAP-18; AKAP-18 {ECO:0000250|UniProtKB:F8VQ58}; AKAP-7 isoform alpha; AKAP-7 isoform gamma; AKAP-7 isoforms alpha and beta; AKAP-7 isoforms delta and gamma; akap7; akap7 {ECO:0000312|RGD:1303071}; PRKA7 isoform alpha; PRKA7 isoform gamma; PRKA7 isoforms alpha/beta; PRKA7 isoforms delta and gamma; protein kinase A anchoring protein 7; Protein kinase A-anchoring protein 7 isoform alpha; Protein kinase A-anchoring protein 7 isoform gamma; Protein kinase A-anchoring protein 7 isoforms alpha/beta; Protein kinase A-anchoring protein 7 isoforms delta and gamma
Gene Aliases: 6430401D08; AI662165; AKAP-18; AKAP15; AKAP18; AKAP18d; AKAP7; BB170514
UniProt ID: (Human) O43687, (Mouse) O55074, (Rat) Q6JP77
Entrez Gene ID: (Human) 9465, (Mouse) 432442, (Rat) 361458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.