Invitrogen
Manufacturer Code:PA577705
Catalog # PIPA577705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RGAKRSRPSKLTED corresponding to amino acid residues 128-141 of rat AKAP150 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-kinase anchor protein 150 kDa; A-Kinase Anchor Protein 150kD AKAP5; A-kinase anchor protein 5; AKAP 150; AKAP-5; cAMP-dependent protein kinase regulatory subunit II high affinity-binding protein; RII-B-binding protein
Gene Aliases: 3526401B18Rik; AKAP 150; AKAP-5; AKAP150; Akap5; Akap79; BB098886; Gm258; P150
UniProt ID: (Mouse) D3YVF0
Entrez Gene ID: (Mouse) 238276, (Rat) 171026
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.