Invitrogen
Manufacturer Code:PA536155
Catalog # PIPA536155
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N-terminal of human AKAP 5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 5; A kinase (PRKA) anchor protein 5 A kinase anchor protein 5 A kinase anchor protein 79kDa A-kinase anchor protein 5 A-kinase anchor protein 79 kDa AKAP 5 AKAP 79 AKAP-5 AKAP5 AKAP75 AKAP79; A-kinase anchor protein 150 kDa; A-kinase anchor protein 5; A-kinase anchor protein 79 kDa; A-kinase anchoring protein 75/79; AKAP 150; AKAP 79; AKAP-5; cAMP-dependent protein kinase regulatory subunit II high affinity-binding protein; H21; RII-B-binding protein
Gene Aliases: 3526401B18Rik; AKAP 150; AKAP-5; AKAP150; AKAP5; AKAP75; AKAP79; BB098886; Gm258; H21; P150
UniProt ID: (Human) P24588, (Mouse) D3YVF0
Entrez Gene ID: (Human) 9495, (Mouse) 238276, (Rat) 171026
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.