Invitrogen
Manufacturer Code:PA5101095
Catalog # PIPA5101095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AKAP5 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3526401B18Rik A kinase (PRKA) anchor protein 5 AKAP 150 AKAP 5 AKAP 79 AKAP150 Akap5 AKAP-5 AKAP75 Akap79 A-kinase anchor protein 150 kDa A-kinase anchor protein 5 A-kinase anchor protein 79 kDa A-kinase anchoring protein 5 A-kinase anchoring protein 75/79 BB098886 cAMP-dependent protein kinase regulatory subunit II high affinity-binding protein Gm258 H21 P150 RII-B-binding protein; A kinase (PRKA) anchor protein 5; A-kinase anchor protein 150 kDa; A-kinase anchor protein 5; A-kinase anchor protein 79 kDa; A-kinase anchoring protein 75/79; AKAP 150; AKAP 79; AKAP-5; cAMP-dependent protein kinase regulatory subunit II high affinity-binding protein; H21
Gene Aliases: 3526401B18Rik; AKAP 150; AKAP-5; AKAP150; AKAP5; AKAP75; AKAP79; BB098886; Gm258; H21; P150
UniProt ID: (Human) P24588, (Mouse) D3YVF0
Entrez Gene ID: (Human) 9495, (Mouse) 238276
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.