Abnova
Manufacturer Code:PAB17356
Catalog # 89-119-012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human AKAP5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of AKAP5.
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 5; A-kinase anchor protein 5; A-kinase anchor protein 79 kDa; A-kinase anchoring protein 75/79; AKAP 79; AKAP-5; AKAP75; AKAP79; cAMP-dependent protein kinase regulatory subunit II high affinity-binding protein; H21
Gene Aliases: AKAP5; AKAP75; AKAP79; H21
UniProt ID: (Human) P24588
Entrez Gene ID: (Human) 9495
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.