Proteintech
Manufacturer Code:139071AP150UL
Catalog # 50-172-9083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AKAP3 Fusion Protein Ag4885 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 3; A kinase anchor protein 3 AKAP 110 AKAP 3 AKAP110 AKAP3 Cancer/testis antigen 82 CT82 Fibrousheathin 1 Fibrousheathin I FSP95 PRKA3 SOB1 Sperm oocyte binding protein; A-kinase anchor protein 110 kDa; A-kinase anchor protein 3; A-kinase anchor protein, 110kDa; AKAP 110; AKAP-3; Cancer/testis antigen 82; CT82; epididymis luminal protein 159; Fibrous Sheath Protein of 95 kDa; Fibrousheathin I; Fibrousheathin-1; FSP95; PRKA3; protein kinase A binding protein AKAP 110; Protein kinase A-anchoring protein 3; Sperm oocyte-binding protein; sperm oocyte-binding protein 1
Gene Aliases: AKAP 110; AKAP110; AKAP3; CT82; FSP95; HEL159; PRKA3; SOB1
UniProt ID: (Human) O75969, (Mouse) O88987
Entrez Gene ID: (Human) 10566, (Mouse) 11642, (Rat) 312720
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.