Proteintech
Manufacturer Code:146221AP150UL
Catalog # 50-172-9592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AKAP14 Fusion Protein Ag6255 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 14; A kinase anchor protein 14 A kinase anchor protein 28 kDa AKAP 14 AKAP 28 AKAP14 AKAP28; A-kinase anchor protein 14; A-kinase anchor protein 28 kDa; A-kinase anchoring protein 28; AKAP 28; AKAP-14; PRKA14; Protein kinase A-anchoring protein 14
Gene Aliases: AKAP14; AKAP28; PRKA14
UniProt ID: (Human) Q86UN6
Entrez Gene ID: (Human) 158798
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.