Invitrogen
Manufacturer Code:PA536342
Catalog # PIPA536342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 700-750 of human AKAP 13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 13; A-kinase anchor protein 13; A-kinase anchor protein 13 AKAP-13 AKAP-Lbc Breast cancer nuclear receptor-binding auxiliary protein Guanine nucleotide exchange factor Lbc Human thyroid-anchoring protein 31 Lymphoid blast crisis oncogene LBC oncogene Non-oncogenic Rho GTPase-specific GTP exchange factor Protein kinase A-anchoring protein 13 PRKA13 p47 BRX HT31 LBC AKAP13; A-kinase anchoring protein; AKAP-13; AKAP-Lbc; Breast cancer nuclear receptor-binding auxiliary protein; Guanine nucleotide exchange factor Lbc; Human thyroid-anchoring protein 31; LBC oncogene; Lymphoid blast crisis oncogene; Non-oncogenic Rho GTPase-specific GTP exchange factor; p47; PRKA13; Protein kinase A-anchoring protein 13
Gene Aliases: AKAP-13; AKAP-Lbc; AKAP13; ARHGEF13; BRX; c-lbc; HA-3; HT31; LBC; p47; PRKA13; PROTO-LB; PROTO-LBC
UniProt ID: (Human) Q12802
Entrez Gene ID: (Human) 11214
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.