Invitrogen
Manufacturer Code:PA5114833
Catalog # PIPA5114833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AKAP12 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein (gravin) 12 A kinase (PRKA) anchor protein 12 AI317366 AKAP 250 AKAP12 AKAP-12 AKAP12A AKAP12B AKAP12G AKAP250 A-kinase anchor protein 12 A-kinase anchor protein 250 kDa A-kinase anchor protein 250kDa A-kinase anchoring protein 12 Gag12 gercelin Germ cell lineage protein gercelin Gravin kinase scaffold protein gravin Myasthenia gravis autoantigen myasthenia gravis autoantigen gravin Srcs5 Src-Suppressed C Kinase Substrate SSeCKS SSeCKS Gravin AKAP250 testis specific gene A12 Tsga12 v-src suppressed transcript 5; A kinase (PRKA) anchor protein 12; A-kinase anchor protein 12; A-kinase anchor protein 250 kDa; A-kinase anchor protein, 250kDa; AKAP 250; AKAP-12; Gravin; kinase scaffold protein gravin; Myasthenia gravis autoantigen; myasthenia gravis autoantigen gravin; Src-Suppressed C Kinase Substrate
Gene Aliases: AKAP12; AKAP250; SSeCKS
UniProt ID: (Human) Q02952
Entrez Gene ID: (Human) 9590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.