Invitrogen
Manufacturer Code:PA5111897
Catalog # PIPA5111897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 1639-1652 of mouse AKAP12 (Intracellular C-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein (gravin) 12; A kinase (PRKA) anchor protein 12; A-kinase anchor protein 12; A-kinase anchor protein 250 kDa; A-kinase anchor protein, 250kDa; AI317366 AKAP 250 Akap12 AKAP-12 AKAP12A AKAP12B AKAP12G AKAP250 A-kinase anchor protein 12 A-kinase anchor protein 250 kDa Gag12 Germ cell lineage protein gercelin Gravin Myasthenia gravis autoantigen Srcs5 Src-suppressed C kinase substrate Ssecks Tsga12; AKAP 250; AKAP-12; gercelin; Germ cell lineage protein gercelin; Gravin; kinase scaffold protein gravin; Myasthenia gravis autoantigen; myasthenia gravis autoantigen gravin; Src-Suppressed C Kinase Substrate; SSeCKS; SSeCKS, Gravin, AKAP250; testis specific gene A12; v-src suppressed transcript 5
Gene Aliases: AI317366; AKAP12; AKAP12A; AKAP12B; AKAP12G; AKAP250; Gag12; Srcs5; SSeCKS; Tsga12
UniProt ID: (Human) Q02952, (Mouse) Q9WTQ5, (Rat) Q5QD51
Entrez Gene ID: (Human) 9590, (Mouse) 83397, (Rat) 83425
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.