Invitrogen
Manufacturer Code:PA129372
Catalog # PIPA129372
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rat SSecks fragment corresponding to amino acid 620-1596. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein (gravin) 12; A-kinase anchor protein 12; AKAP-12; AKAP12A AKAP12B AKAP12G; SSeCKS, Gravin, AKAP250
Gene Aliases: Akap12; AKAP12A; AKAP12B; AKAP12G
UniProt ID: (Rat) Q5QD51
Entrez Gene ID: (Rat) 83425
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.