Proteintech
Manufacturer Code:251991AP150UL
Catalog # 50-173-4781
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AKAP12 Fusion Protein Ag17769 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein (gravin) 12; A kinase (PRKA) anchor protein 12; A kinase anchor protein 12 AKAP 12 AKAP 250 AKAP12 AKAP250 Gravin Myasthenia gravis autoantigen; A-kinase anchor protein 12; A-kinase anchor protein 250 kDa; A-kinase anchor protein, 250kDa; AKAP 250; AKAP-12; Gravin; kinase scaffold protein gravin; Myasthenia gravis autoantigen; myasthenia gravis autoantigen gravin; Src-Suppressed C Kinase Substrate; SSeCKS, Gravin, AKAP250
Gene Aliases: AKAP12; AKAP12A; AKAP12B; AKAP12G; AKAP250; SSeCKS
UniProt ID: (Human) Q02952, (Rat) Q5QD51
Entrez Gene ID: (Human) 9590, (Rat) 83425
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.