Invitrogen
Manufacturer Code:PA585225
Catalog # PIPA585225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human AKAP10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 10; A kinase anchor protein 10; A kinase anchor protein 10, mitochondrial; A-kinase anchor protein 10, mitochondrial; AKAP-10; D-AKAP-2; D-AKAP2 antibody MGC9414 antibody PRKA10 antibody AKAP10 antibody dual specificity A kinase-anchoring protein 2 antibody D-AKAP-2 antibody a kinase anchor protein 10 mitochondrial antibody A-kinase anchor protein 10 mitochondrial antibody dual-specificity A-kinase anchoring protein 2 antibody protein kinase A anchoring protein 10 antibody protein kinase A-anchoring protein 10 antibody AKAP-10 antibody mitochondrial A kinase PPKA anchor protein 10 antibody A kinase (PRKA) anchor protein 10 antibody; Dual specific A kinase anchoring protein 2; Dual specificity A kinase-anchoring protein 2; mitochondrial A kinase PPKA anchor protein 10; PRKA10; protein kinase A anchoring protein 10; Protein kinase A-anchoring protein 10
Gene Aliases: 1500031L16Rik; AKAP-10; AKAP10; B130049N18Rik; D-AKAP-2; D-AKAP2; PRKA10
UniProt ID: (Human) O43572, (Mouse) O88845
Entrez Gene ID: (Human) 11216, (Rat) 360540, (Mouse) 56697
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.