Invitrogen
Manufacturer Code:PA518959
Catalog # PIPA518959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RKVKGKEQEKTSD) corresponding to the internal amino acids of AKAP10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase anchor protein 10, mitochondrial; A-kinase anchor protein 10, mitochondrial; AKAP10 A kinase (PRKA) anchor protein 10 D-AKAP2 MGC9414 PRKA10 A-kinase anchor protein 10 dual-specificity A-kinase anchoring protein 2 mitochondrial A kinase PPKA anchor protein 10 protein kinase A anchoring protein 10; Dual specific A kinase anchoring protein 2
Gene Aliases: D-akap2
Entrez Gene ID: (Rat) 360540
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.