Abnova
Manufacturer Code:PAB7481
Catalog # 89-133-292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human AKAP10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of AKAP10.
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 10; A kinase anchor protein 10; A-kinase anchor protein 10, mitochondrial; AKAP-10; D-AKAP-2; D-AKAP2; Dual specificity A kinase-anchoring protein 2; MGC9414; mitochondrial A kinase PPKA anchor protein 10; PRKA10; protein kinase A anchoring protein 10; Protein kinase A-anchoring protein 10
Gene Aliases: AKAP-10; AKAP10; D-AKAP-2; D-AKAP2; PRKA10
UniProt ID: (Human) O43572
Entrez Gene ID: (Human) 11216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.