Novus Biologicals
Manufacturer Code:NBP271813V
Catalog # NBP271813V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F9 |
Immunogen | Full length recombinant protein of human AK5 (NP_777283) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: adenylate kinase 5 adenylate kinase 6 adenylate kinase isoenzyme 5 AK 5 AK6 ATP-AMP transphosphorylase 5 EC 2.7.4.3 MGC33326; adenylate kinase 6; Adenylate kinase isoenzyme 5; AK 5; ATP-AMP transphosphorylase 5
Gene Aliases: AK5; AK6
UniProt ID: (Human) Q9Y6K8
Entrez Gene ID: (Human) 26289
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.