OriGene
Manufacturer Code:CF503011
Catalog # 50-166-1806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B1 |
Immunogen | Full length human recombit protein of human AK4 produced in HEK293 cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: adenylate kinase 3 alpha-like 1; Adenylate kinase 3-like; adenylate kinase 3-like 1; adenylate kinase 3-like 2; adenylate kinase 3-like {ECO:0000255|HAMAP-Rule:MF_03170}; Adenylate kinase 4, mitochondrial; adenylate kinase 4, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03170}; Adenylate kinase isoenzyme 4; adenylate kinase isoenzyme 4, mitochondrial; AK 4; AK 4 AK3 AK3L1 AK3L2; AK 4 {ECO:0000255|HAMAP-Rule:MF_03170}; AK4; ATP-AMP transphosphorylase; GTP:AMP phosphotransferase AK4; GTP:AMP phosphotransferase AK4 {ECO:0000255|HAMAP-Rule:MF_03170}; GTP:AMP phosphotransferase AK4, mitochondrial; mitochondrial adenylate kinase-3; nucleoside-triphosphate-adenylate kinase
Gene Aliases: AK 4; Ak-3; Ak-4; AK3; Ak3b; AK3L1; AK3L2; AK4; D4Ertd274e
UniProt ID: (Human) P27144, (Rat) Q9WUS0, (Mouse) Q9WUR9
Entrez Gene ID: (Human) 205, (Dog) 489554, (Rat) 29223, (Mouse) 11639
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.