Proteintech
Manufacturer Code:125621AP150UL
Catalog # 50-172-8069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AK3 Fusion Protein Ag3146 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate kinase 3; adenylate kinase 3 AK 3 AK3 AK3L1 AK6 AKL3L AKL3L1 FIX; Adenylate kinase 3 alpha-like 1; adenylate kinase 3 alpha-like 1 {ECO:0000255|HAMAP-Rule:MF_03169}; adenylate kinase 3 {ECO:0000255|HAMAP-Rule:MF_03169}; adenylate kinase 6, adenylate kinase 3 like 1; AK 3; AK 3 {ECO:0000255|HAMAP-Rule:MF_03169}; AK3; GTP:AMP phosphotransferase AK3, mitochondrial; GTP:AMP phosphotransferase AK3, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03169}; GTP:AMP phosphotransferase mitochondrial; GTP:AMP phosphotransferase, mitochondrial
Gene Aliases: AA407498; AI506714; AK-3; AK3; Ak3l; AK3L1; AK6; AKL3L; AKL3L1; FIX
UniProt ID: (Human) Q9UIJ7, (Rat) P29411, (Mouse) Q9WTP7
Entrez Gene ID: (Human) 50808, (Rat) 26956, (Mouse) 56248
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.