Proteintech
Manufacturer Code:149781AP150UL
Catalog # 50-172-9881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AK1 Fusion Protein Ag6890 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylate kinase 1 Adenylate kinase isoenzyme 1 AK 1 AK1 ATP AMP transphosphorylase 1 Myokinase; Adenylate kinase isoenzyme 1; adenylate kinase isoenzyme 1 {ECO:0000255|HAMAP-Rule:MF_03171}; Adenylate monophosphate kinase; adenylate monophosphate kinase {ECO:0000255|HAMAP-Rule:MF_03171}; AK 1; AK 1 {ECO:0000255|HAMAP-Rule:MF_03171}; ATP-AMP transphosphorylase 1; ATP-AMP transphosphorylase 1 {ECO:0000255|HAMAP-Rule:MF_03171}; ATP:AMP phosphotransferase; ATP:AMP phosphotransferase {ECO:0000255|HAMAP-Rule:MF_03171}; cytosolic adenylate kinase; Myokinase; myokinase {ECO:0000255|HAMAP-Rule:MF_03171}; testis secretory sperm binding protein Li 58j
Gene Aliases: Ak 1; Ak-1; AK1; B430205N08Rik; HTL-S-58j
UniProt ID: (Human) P00568, (Mouse) Q9R0Y5, (Rat) P39069
Entrez Gene ID: (Human) 203, (Mouse) 11636, (Rat) 24183
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.