Invitrogen
Manufacturer Code:PA5109373
Catalog # PIPA5109373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein of human AJUBA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ajuba homolog; AJUBA JUB LIM domain-containing protein ajuba; Ajuba protein; jub, ajuba homolog; LIM domain-containing protein ajuba; protein ajuba
Gene Aliases: AJUBA; JUB
UniProt ID: (Human) Q96IF1, (Rat) Q5U2Z2
Entrez Gene ID: (Human) 84962, (Rat) 85265
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.