Invitrogen
Manufacturer Code:PA518525
Catalog # PIPA518525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KDVDLSQPRKGRKP) corresponding to the internal amino acids of AIRE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AIRE autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy) AIRE1 APECED APS1 APSI PGA1 autoimmune regulator (APECED protein) autoimmune regulator (automimmune polyendocrinopathy candidiasis ectodermal dystrophy) autoimmune regulator AIRE; APECED protein; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; Autoimmune regulator
Gene Aliases: AIRE; AIRE1; APECED; APS1; APSI; PGA1
UniProt ID: (Human) O43918
Entrez Gene ID: (Human) 326, (Pig) 100621659
Molecular Function: ligase transcription cofactor transcription factor ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.