Abnova
Manufacturer Code:PAB7040
Catalog # 89-132-899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human AIRE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of AIRE.
For Research Use Only
Protein Aliases: AIRE1; APECED; APECED protein; APS1; APSI; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; Autoimmune regulator; PGA1
Gene Aliases: AIRE; AIRE1; APECED; APS1; APSI; PGA1
UniProt ID: (Human) O43918
Entrez Gene ID: (Human) 326
Molecular Function: ligase transcription cofactor transcription factor ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.