Invitrogen
Manufacturer Code:PA542294
Catalog # PIPA542294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human WWP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIP2; atrophin-1 interacting protein 2; Atrophin-1-interacting protein 2; HECT-type E3 ubiquitin transferase WWP2; NEDD4-like E3 ubiquitin-protein ligase WWP2; WW domain containing E3 ubiquitin protein ligase 2 AIP2 WWp2-like; WW domain-containing protein 2; WW domain-containing protein 4
Gene Aliases: 1300010O06Rik; AA690238; AIP2; AW554328; id:ibd1121; wu:fo87e10; WWP2; WWp2-like; zgc:154036
UniProt ID: (Human) F5H213, (Mouse) Q9DBH0
Entrez Gene ID: (Sheep) 101122376, (Rabbit) 100344222, (Dog) 479672, (Human) 11060, (Horse) 100067291, (Bovine) 512457, (Rat) 291999, (Guinea pig) 100721341, (Mouse) 66894, (Zebrafish) 564527
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.