OriGene
Manufacturer Code:CF505031
Catalog # 50-166-3328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B6 |
Immunogen | Full length human recombit protein of human AIP produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AH receptor-interacting protein; AIP; ARA9 FKBP16 FKBP37 PITA1 SMTPHN XAP-2 XAP2; Aryl-hydrocarbon receptor-interacting protein; HBV X-associated protein 2; Immunophilin homolog ARA9; XAP-2
Gene Aliases: AIP; ARA9; FKBP16; FKBP37; SMTPHN; XAP-2; XAP2
UniProt ID: (Human) O00170
Entrez Gene ID: (Human) 9049
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.