Invitrogen
Manufacturer Code:MA316515
Catalog # AMSA6515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 35-2 |
Immunogen | Bacterially expressed human ARA9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AH receptor-interacting protein; AIP; AIP Aryl Hydrocarbon Receptor Interacting Protein FKBP37 HBVX Associated Protein XAP2; Aryl-hydrocarbon receptor-interacting protein; HBV X-associated protein 2; HBV-X associated protein; Immunophilin homolog ARA9; Immunophilin XAP2; XAP-2
Gene Aliases: AA408703; AIP; ARA9; AW476050; D19Bwg1412e; FKBP16; FKBP37; SMTPHN; XAP-2; XAP2
UniProt ID: (Human) O00170, (Mouse) O08915, (Rat) Q5FWY5
Entrez Gene ID: (Human) 9049, (Mouse) 11632, (Rat) 282827
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.