Abnova
Manufacturer Code:MAB2382
Catalog # 89-110-977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 35-2 |
Immunogen | Recombinant protein corresponding to human AIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant AIP.
For Research Use Only
Protein Aliases: AH receptor-interacting protein; AIP; ARA9; Aryl-hydrocarbon receptor-interacting protein; FKBP16; FKBP37; HBV X-associated protein 2; Immunophilin homolog ARA9; SMTPHN; XAP-2; XAP2
Gene Aliases: AIP; ARA9; FKBP16; FKBP37; SMTPHN; XAP-2; XAP2
UniProt ID: (Human) O00170
Entrez Gene ID: (Human) 9049
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.