Novus Biologicals
Manufacturer Code:NBP222451
Catalog # NBP222451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length bacterially expressed mouse recombinant AIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: AH receptor-interacting protein; AIP; ARA9 FKBP16 FKBP37 SMTPHN XAP2 XAP-2; Aryl-hydrocarbon receptor-interacting protein; HBV X-associated protein 2; Immunophilin homolog ARA9; XAP-2
Gene Aliases: AIP; ARA9; FKBP16; FKBP37; SMTPHN; XAP-2; XAP2
UniProt ID: (Human) O00170
Entrez Gene ID: (Human) 9049
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.