Novus Biologicals
Manufacturer Code:NBP146149
Catalog # NBP146149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TARHFHTEERLSTPH corresponding to internal region according to NP_001018070.1 NP_001139760.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AIFLFLJ30473 apoptosis-inducing factor 3 apoptosis-inducing factor like apoptosis-inducing factor mitochondrion-associated 3 Apoptosis-inducing factor-like protein FLJ45137; Apoptosis-inducing factor 3; apoptosis-inducing factor like; apoptosis-inducing factor, mitochondrion-associated, 3; Apoptosis-inducing factor-like protein
Gene Aliases: AIFL; AIFM3
UniProt ID: (Human) Q96NN9
Entrez Gene ID: (Human) 150209
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.