Abnova
Manufacturer Code:H00084883K
Catalog # 89-200-690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human AIFM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human AIFM2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: AMID; AMID PRG3 RP11-367H5.2; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated, 2; Ferroptosis suppressor protein 1; FSP1; p53-responsive gene 3 protein
Gene Aliases: AIFM2; AMID; PRG3
UniProt ID: (Human) Q9BRQ8
Entrez Gene ID: (Human) 84883
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.