OriGene
Manufacturer Code:CF811161
Catalog # 50-167-0483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4E6 |
Immunogen | Human recombit protein fragment corresponding to amino acids 134-483 of human AIFM1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AIF CMT2D CMTX4 COWCK COXPD6 DFNX5 NADMR NAMSD PDCD8; Apoptosis-inducing factor 1, mitochondrial; apoptosis-inducing factor, mitochondrion-associated, 1; harlequin; programmed cell death 8 (apoptosis-inducing factor); Programmed cell death protein 8; striatal apoptosis-inducing factor; testicular secretory protein Li 4
Gene Aliases: AIF; AIFM1; AIFsh2; CMT2D; CMTX4; COWCK; COXPD6; DFNX5; Hq; NADMR; NAMSD; PDCD8
UniProt ID: (Human) O95831, (Mouse) Q9Z0X1
Entrez Gene ID: (Human) 9131, (Mouse) 26926
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.