Invitrogen
Manufacturer Code:MA515880
Catalog # PIMA515880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4E7E11 |
Immunogen | Purified recombit fragment of human AIF expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis-inducing factor 1, mitochondrial; apoptosis-inducing factor, mitochondrion-associated, 1; harlequin; programmed cell death 8 (apoptosis-inducing factor); Programmed cell death protein 8; RP3-438D16.2 AIF COXPD6 PDCD8 apoptosis-inducing factor 1 mitochondrial programmed cell death 8 (apoptosis-inducing factor) striatal apoptosis-inducing factor; striatal apoptosis-inducing factor; testicular secretory protein Li 4
Gene Aliases: AIF; AIFM1; AIFsh2; CMT2D; CMTX4; COWCK; COXPD6; DFNX5; Hq; NADMR; NAMSD; PDCD8
UniProt ID: (Human) O95831, (Mouse) Q9Z0X1, (Rat) Q9JM53
Entrez Gene ID: (Human) 9131, (Mouse) 26926, (Rat) 83533
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.