Novus Biologicals
Manufacturer Code:NB1002833
Catalog # NB1002833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | This AIF-1/Iba1 Antibody was developed against a peptide with sequence C-NKQFLDDPKYSSDED corresponding to internal region according to NP_001614.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AIF-1; AIF-1 allograft inflammatory factor 1 G1 IBA1 IbaI interferon gamma responsive transcript Ionized calcium-binding adapter molecule 1 IRT1 IRT-1 Protein G1; Allograft inflammatory factor 1; interferon gamma responsive transcript; Ionized calcium-binding adapter molecule 1; Protein G1
Gene Aliases: AIF-1; AIF1; G1; IBA1; IRT-1; IRT1
UniProt ID: (Human) P55008
Entrez Gene ID: (Human) 199
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.