Invitrogen
Manufacturer Code:14595937
Catalog # 14-595-937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | mAID-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: activation induced deaminase; activation induced deaminase activation-induced cytidine deaminase AICDA AID Arp2 CDA2 cytidine aminohydrolase epididymis secretory protein Li 284 HEL-S-284 HIGM2 integrated into Burkitt's lymphoma cell line Ramos single-stranded DNA cytosine deaminase; Activation-induced cytidine deaminase; AID; Cytidine aminohydrolase; epididymis secretory protein Li 284; integrated into Burkitt's lymphoma cell line Ramos; Single-stranded DNA cytosine deaminase
Gene Aliases: AICDA; AID; ARP2; CDA2; HEL-S-284; HIGM2
UniProt ID: (Human) Q9GZX7, (Mouse) Q9WVE0
Entrez Gene ID: (Human) 57379, (Mouse) 11628
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.