Invitrogen
Manufacturer Code:PA534918
Catalog # PIPA534918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contains a sequence corresponding to a region within amino acids 67 and 228 of AICDA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activation-induced cytidine deaminase; AID; AID ARP2 CDA2 HIGM2; Cytidine aminohydrolase; epididymis secretory protein Li 284; integrated into Burkitt's lymphoma cell line Ramos; Single-stranded DNA cytosine deaminase
Gene Aliases: AICDA; AID; ARP2; CDA2; HEL-S-284; HIGM2
UniProt ID: (Human) Q9GZX7
Entrez Gene ID: (Human) 57379
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.