OriGene
Manufacturer Code:TA504670
Catalog # 50-167-5309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H2 |
Immunogen | Full length human recombit protein of human AHSG produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A2HS AHS APMR1 FETUA HSGA; Alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; Ba-alpha-2-glycoprotein; fetuin A; Fetuin-A
Gene Aliases: A2HS; AHS; AHSG; FETUA; HSGA; PRO2743
UniProt ID: (Human) P02765
Entrez Gene ID: (Human) 197
Molecular Function:
cysteine protease inhibitor
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.