Novus Biologicals
Manufacturer Code:NBP178205
Catalog # NBP178205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 25 and 75 of human Abelson Helper Integration site 1 using the numbering given in entry NP_001128302.1 (GeneID 54806). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson helper integration site 1 protein homolog; Abelson helper integration site Abelson helper integration site 1 Abelson helper integration site 1 protein homolog AHI-1 contatins SH3 and WD40 domains dJ71N10.1 DKFZp686J1653 FLJ14023 FLJ20069 JBTS3 jouberin ORF1; AHI-1; contatins SH3 and WD40 domains; Jouberin
Gene Aliases: AHI-1; AHI1; dJ71N10.1; JBTS3; ORF1
UniProt ID: (Human) Q9H0H2
Entrez Gene ID: (Human) 54806
Molecular Function:
RNA binding protein
enzyme modulator
esterase
hydrolase
kinase inhibitor
kinase modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.