Abnova
Manufacturer Code:PAB6699
Catalog # 89-132-612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human AHI1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of AHI1.
For Research Use Only
Protein Aliases: Abelson helper integration site 1 protein homolog; AHI-1; contatins SH3 and WD40 domains; dJ71N10.1; DKFZp686J1653; FLJ14023; FLJ20069; JBTS3; Jouberin; ORF1
Gene Aliases: AHI-1; AHI1; dJ71N10.1; JBTS3; ORF1
UniProt ID: (Human) Q9H0H2
Entrez Gene ID: (Human) 54806
Molecular Function:
RNA binding protein
enzyme modulator
esterase
hydrolase
kinase inhibitor
kinase modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.