Abnova
Manufacturer Code:H00000191M09A
Catalog # 89-104-058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M2 |
Immunogen | AHCY (AAH116061 a.a. &bsim 432 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant AHCY.
For Research Use Only
Protein Aliases: Adenosylhomocysteinase; AdoHcyase; S-adenosyl-L-homocysteine hydrolase; S-adenosylhomocysteine hydrolase; SAHH
Gene Aliases: adoHcyase; AHCY; SAHH
UniProt ID: (Human) P23526
Entrez Gene ID: (Human) 191
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.