Invitrogen
Manufacturer Code:PA576705
Catalog # PIPA576705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human AHCY. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenosylhomocysteinase; AdenosylhomocysteinaseAdoHcyaseahcYS adenosyl L homocysteine hydrolaseS adenosylhomocysteine hydrolaseS-adenosyl-L-homocysteine hydrolaseSAHHSAHH&lowbarHUMAN; AdoHcyase; CUBP; Liver copper-binding protein; S-adenosyl-L-homocysteine hydrolase; S-adenosylhomocysteine hydrolase
Gene Aliases: AA987153; adoHcyase; AHCY; AL024110; CuBP; SAHH
UniProt ID: (Human) P23526, (Mouse) P50247
Entrez Gene ID: (Human) 191, (Mouse) 269378
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.