Invitrogen
Manufacturer Code:PA569791
Catalog # PIPA569791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human AHCY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3D-structure Acetylation Cytoplasm Direct protein sequencing Hydrolase NAD One-carbon metabolism Polymorphism.; Adenosylhomocysteinase; AdoHcyase; CUBP; dtp; Liver copper-binding protein; S-adenosyl-L-homocysteine hydrolase; S-adenosylhomocysteine hydrolase
Gene Aliases: AA987153; adoHcyase; AHCY; AL024110; cb1079; CuBP; D150; ducttrip; hm:zeh1173; hm:zeh1364; SAHH; wu:fj67b02
UniProt ID: (Bovine) Q3MHL4, (Human) P23526, (Mouse) P50247, (Rat) P10760
Entrez Gene ID: (Rabbit) 100347222, (Bovine) 508158, (Dog) 477198, (Goat) 102191350, (Human) 191, (Mouse) 269378, (Rat) 29443, (Guinea pig) 100723355, (Zebrafish) 387530
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.