Invitrogen
Manufacturer Code:PA512311
Catalog # PIPA512311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 79-110 amino acids from the N-terminal region of human S adenosylhomocysteine hydrolase (ACHY) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3D-structure Acetylation Cytoplasm Direct protein sequencing Hydrolase NAD One-carbon metabolism Polymorphism.; Adenosylhomocysteinase; AdoHcyase; S-adenosyl-L-homocysteine hydrolase; S-adenosylhomocysteine hydrolase
Gene Aliases: adoHcyase; AHCY; SAHH
UniProt ID: (Human) P23526
Entrez Gene ID: (Human) 191
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.