Invitrogen
Manufacturer Code:PA5109371
Catalog # PIPA5109371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 253-432 of human AHCY (NP_000678.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA987153 Acetylation Adenosylhomocysteinase AdoHcyase AHCY AL024110 BOS_13353 cb1079 CUBP D150 ducttrip hm:zeh1173 hm:zeh1364 Hydrolase Liver copper-binding protein NAD Polymorphism S-adenosyl-L-homocysteine hydrolase SAHH wu:fj67b02; Adenosylhomocysteinase; AdoHcyase; S-adenosyl-L-homocysteine hydrolase; S-adenosylhomocysteine hydrolase
Gene Aliases: adoHcyase; AHCY; SAHH
UniProt ID: (Human) P23526, (Rat) P10760
Entrez Gene ID: (Human) 191, (Rat) 29443
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.