Invitrogen
Manufacturer Code:PA5102718
Catalog # PIPA5102718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AHCY corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA987153 Acetylation Adenosylhomocysteinase adoHcyase AHCY AL024110 CuBP Hydrolase Liver copper-binding protein NAD Polymorphism S-adenosylhomocysteine hydrolase S-adenosyl-L-homocysteine hydrolase SAHH; Adenosylhomocysteinase; AdoHcyase; CUBP; Liver copper-binding protein; S-adenosyl-L-homocysteine hydrolase; S-adenosylhomocysteine hydrolase
Gene Aliases: AA987153; adoHcyase; AHCY; AL024110; CuBP; SAHH
UniProt ID: (Human) P23526, (Mouse) P50247, (Rat) P10760
Entrez Gene ID: (Human) 191, (Mouse) 269378, (Rat) 29443
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.