Proteintech
Manufacturer Code:666021IG150UL
Catalog # 50-173-7020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A5B12 |
Immunogen | AGXT2 Fusion Protein Ag26888 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (R)-3-amino-2-methylpropionate--pyruvate transaminase; AGT 2; AGT 2 AGT2 AGXT2 Beta ALAAT II D AIBAT; Alanine--glyoxylate aminotransferase 2, mitochondrial; Beta-ALAAT II; Beta-alanine-pyruvate aminotransferase; D-AIBAT
Gene Aliases: AGT2; AGXT2; AI303810; AI663818; DAIBAT
UniProt ID: (Human) Q9BYV1, (Mouse) Q3UEG6
Entrez Gene ID: (Human) 64902, (Mouse) 268782
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.