Invitrogen
Manufacturer Code:PA543259
Catalog # PIPA543259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human AGXT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (R)-3-amino-2-methylpropionate--pyruvate transaminase; AGT 2; Alanine--glyoxylate aminotransferase 2, mitochondrial; Beta-ALAAT II; Beta-alanine-pyruvate aminotransferase; D-AIBAT
Gene Aliases: AGT2; AGXT2; AI303810; AI663818; DAIBAT
UniProt ID: (Bovine) Q17QF0, (Human) Q9BYV1, (Mouse) Q3UEG6, (Rat) Q64565
Entrez Gene ID: (Bovine) 521553, (Horse) 100053849, (Human) 64902, (Rabbit) 100352336, (Dog) 612589, (Mouse) 268782, (Rat) 83784
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.