Invitrogen
Manufacturer Code:PA526833
Catalog # PIPA526833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 437-463 amino acids from the C-terminal region of human AGXT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (R)-3-amino-2-methylpropionate--pyruvate transaminase; AGT 2; AGT2 DAIBAT (R)-3-amino-2-methylpropionate--pyruvate transaminase alanine--glyoxylate aminotransferase 2 mitochondrial beta-ALAAT II beta-alanine-pyruvate aminotransferase; Alanine--glyoxylate aminotransferase 2, mitochondrial; Beta-ALAAT II; Beta-alanine-pyruvate aminotransferase; D-AIBAT
Gene Aliases: AGT2; AGXT2; DAIBAT
UniProt ID: (Human) Q9BYV1
Entrez Gene ID: (Human) 64902
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.