Novus Biologicals
Manufacturer Code:NBP179303
Catalog # NBP179303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human AGXT2The immunogen for this antibody is AGXT2. Peptide sequence TSVTKLSLHTKPRMPPCDFMPERYQSLGYNRVLEIHKEHLSPVVTAYFQK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (R)-3-amino-2-methylpropionate--pyruvate transaminase; (R)-3-amino-2-methylpropionate--pyruvate transaminase AGT 2 alanine-glyoxylate aminotransferase 2 alanine--glyoxylate aminotransferase 2 Beta-ALAAT II Beta-alanine-pyruvate aminotransferase DAIBAT D-AIBAT EC 2.6.1.40 EC 2.6.1.44 mitochondrial; AGT 2; Alanine--glyoxylate aminotransferase 2, mitochondrial; Beta-ALAAT II; Beta-alanine-pyruvate aminotransferase; D-AIBAT
Gene Aliases: AGT2; AGXT2; DAIBAT
UniProt ID: (Human) Q9BYV1
Entrez Gene ID: (Human) 64902
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.