Novus Biologicals
Manufacturer Code:NBP144376UV
Catalog # NBP144376UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2T4 |
Immunogen | Recombinant AGXT (330-392aa) purified from E. coli |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AGT; AGT1L-alanine: glyoxylate aminotransferase 1 AGThepatic peroxisomal alanine:glyoxylate aminotransferase AGXT1 alanine-glyoxylate aminotransferase Alanine--glyoxylate aminotransferase EC 2.6.1.44 EC 2.6.1.51 PH1 serine:pyruvate aminotransferase SPATserine-pyruvate aminotransferase SPTserine--pyruvate aminotransferase TLH6; Alanine--glyoxylate aminotransferase; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; Serine--pyruvate aminotransferase; serine-pyruvate aminotransferase; serine:pyruvate aminotransferase; SPT
Gene Aliases: AGT; AGT1; AGXT; AGXT1; PH1; SPAT; SPT; TLH6
UniProt ID: (Human) P21549
Entrez Gene ID: (Human) 189
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.