Invitrogen
Manufacturer Code:MA537725
Catalog # PIMA537725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1739CT804.82.83.1 |
Immunogen | recombinant protein between 188-375 amino acids from the human AGXT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AGT; AGT AGT1 AGXT AGXT1 alanine-glyoxylate aminotransferase Alanine--glyoxylate aminotransferase alanine-glyoxylate aminotransferase 1 angiotensin receptor 2 hepatic peroxisomal alanine:glyoxylate aminotransferase L-alanine: glyoxylate aminotransferase 1 PH1 serine:pyruvate aminotransferase serine:pyruvate aminotransferase SPT serine:pyruvate/alanine:glyoxylate aminotransferase serine-pyruvate aminotransferase Serine--pyruvate aminotransferase serine--pyruvate aminotransferase mitochondrial serine--pyruvate aminotransferase mitochondrial serine--pyruvate aminotransferase peroxisomal serine--pyruvate aminotransferase peroxisomal serine--pyruvate aminotransferase serine--pyruvate aminotransferase mitochondrial serine--pyruvate aminotransferase peroxisomal SPAT SPT TLH6; Alanine--glyoxylate aminotransferase; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; Serine--pyruvate aminotransferase; serine-pyruvate aminotransferase; serine:pyruvate aminotransferase; SPT
Gene Aliases: AGT; AGT1; AGXT; AGXT1; PH1; SPAT; SPT; TLH6
UniProt ID: (Human) P21549
Entrez Gene ID: (Human) 189
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.