Abnova
Manufacturer Code:PAB15605
Catalog # 89-117-677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at internal region of human AGXT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised aganist synthetic peptide of AGXT.
For Research Use Only
Protein Aliases: AGT; AGT1; AGXT1; Alanine--glyoxylate aminotransferase; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; PH1; Serine--pyruvate aminotransferase; serine-pyruvate aminotransferase; serine:pyruvate aminotransferase; SPAT; SPT; TLH6
Gene Aliases: AGT; AGT1; AGXT; AGXT1; PH1; SPAT; SPT; TLH6
UniProt ID: (Human) P21549
Entrez Gene ID: (Human) 189
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.